Learn More
Medchemexpress LLC Pyrroloquinoline quinone disodium salt | 122628-50-6 | 98.0% | 374.17 g/mol | C14H4N2Na2O8 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY100196A25MG
Encompass_Preferred
Pyrroloquinoline quinone disodium salt is a redox cofactor used in biochemical and cell biology research. Supplied as the disodium salt with high purity, it is intended for studies of enzymatic activity, mitochondrial function, and oxidative stress.
  • High purity (98.0%) for research-grade experiments.
  • Redox cofactor useful in enzymology and metabolic studies.
  • Suitable for cellular assays examining mitochondrial function and oxidative stress.
  • Disodium salt form improves aqueous solubility compared with the free acid.
  • Provided in small milligram pack sizes suitable for preliminary and in vitro work.
Catalog No. 50-002-15831