Learn More
Medchemexpress LLC Pyruvate Carboxylase-IN-5 | 3047101-14-1 | 98.9% | 294.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY163771100MG
Encompass_Preferred
Pyruvate Carboxylase-IN-5 (compound 6m) is a highly selective and permeable inhibitor of pyruvate carboxylase. This mitochondrial enzyme catalyzes the carboxylation of pyruvate to oxaloacetate, a crucial process for maintaining steady-state levels of Krebs cycle intermediates, which serve as precursors for the synthesis of biomacromolecules like amino acids, fatty acids, and glucose.
  • Pyruvate carboxylase inhibitor
  • High selectivity
  • High permeability
  • For research use only
Catalog No. 50-003-90783