Learn More
Medchemexpress LLC Pyruvate Carboxylase-IN-1 | 70205-50-4 | 99.4% | C15H14O3 | 1 MG
Supplier: Medchemexpress LLC HY1426841MG
Pyruvate Carboxylase-IN-1 is a potent inhibitor of pyruvate carboxylase with antiproliferative activity. It shows inhibitory effects on the growth of HepG2 and HCCLM3 cells.
- Potent pyruvate carboxylase inhibitor
- Exhibits antiproliferative activity
- Inhibits HepG2 cell growth (IC50 1.741 μM)
- Inhibits HCCLM3 cell growth (IC50 8.540 μM)
- For research use only