Learn More
Medchemexpress LLC Pyruvate Carboxylase-IN-5 | 3047101-14-1 | 98.9% | 294.24 | 50 MG
Supplier: Medchemexpress LLC HY16377150MG
Pyruvate Carboxylase-IN-5 (compound 6m) is a pyruvate carboxylase inhibitor with high selectivity and permeability. Pyruvate carboxylase is a mitochondrial enzyme that catalyzes the carboxylation of pyruvate to oxaloacetate, a process that plays an important role in maintaining steady-state levels of Krebs cycle intermediates, which are precursors for the synthesis of biomacromolecules such as amino acids, fatty acids, and glucose.
- High selectivity
- High permeability
- Acts as a pyruvate carboxylase inhibitor