Learn More
Medchemexpress LLC Pyruvate decarboxylase (PDC) | 9001-04-1 | ≥1 U/mg solid | 100 U
Supplier: Medchemexpress LLC HYP2796100U
Pyruvate decarboxylase (PDC) is an enzyme that catalyzes the decarboxylation of pyruvic acid to acetaldehyde. It facilitates the non-oxidative conversion of pyruvate to acetaldehyde and CO2. This enzyme is crucial for alcoholic fermentation and is present in yeast, certain bacteria, and various plants.
- Catalyzes decarboxylation of pyruvic acid to acetaldehyde.
- Facilitates non-oxidative conversion of pyruvate to acetaldehyde and CO2.
- Key enzyme in alcoholic fermentation.
- Found in yeast, bacteria, and plants.
- Subunits are approximately 60 kDa.