Learn More
Bioassay Systems QuantiQuik™ Pyruvate Quick Test Strips. For rapid, direct determination of pyruvate concentrations in food and beverage samples as well as biological samples.
SDP
Supplier:  Bioassay Systems QQPYR10
Encompass_Preferred
For rapid, direct determination of pyruvate concentrations in food and beverage samples as well as biological samples. Key Features: Fast and sensitive. Use of 50, 100, or 400 µL sample. Semi-quantitative measurement between 0-131 mg/L (undiluted) Pyruvate. Convenient. No expensive plate or cuvette readers needed. Sample trea™ent and assay can be performed in under 15 minutes. Method: Visual. Samples: Wine, beer, biological samples, etc. Species: All. Procedure: Assay takes approximately 15 min. Kit size: 10 tests.
Catalog No. 50-225-5895