Learn More
AdipoGen PyTC2
SDP
Supplier:  AdipoGen CDXD0050M100
Encompass_Preferred
Chemical. CAS 3224-36-0. Formula C19H9Cl2N3. MW 350.2. Synthetic. Fluorescent probe with high quantum efficiency, exhibiting single exponential decay kinetics. The radiative fluorescence lifetime is almost constant and independent on molecular surroundings allowing this probe to be utilized in a range of microsurroundings with exciting radiation at a constant wavelength in the blue spectral range. Its fluorescence emission demonstrates a large solvatochromic shift which changes with solvent polarity. For example, the maximum shifts from 435 nm in low polarity n-heptane to 492 nm in high polarity acetonitrile. This product is only sparingly soluble in water
Catalog No. 50-256-1276