Learn More
Medchemexpress LLC PZ703b hydrochloride | 99.2% | 1636.90 g/mol | C80H103Cl2F3N10O11S4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY115718B5MG
Encompass_Preferred
PZ703b hydrochloride is a Bcl-xL-targeting PROTAC degrader that induces apoptosis and inhibits cancer cell proliferation. Supplied as a high-purity hydrochloride salt in small-quantity research packs, it is intended for in vitro studies such as bladder cancer research.
  • Targets Bcl-xL to promote proteasomal degradation and trigger apoptosis.
  • High purity (≈99.2%) suitable for biological assays.
  • Available in small pack sizes for early-stage research.
  • Provided as a hydrochloride salt to aid solubility and handling.
  • Relevant for studies of cancer cell proliferation and bladder cancer models.
Catalog No. 50-002-64670