Learn More
World Precision Instrument PZMIII Binocular Body, pair of 10x eyepieces and eyeguards
SDP
Supplier:  World Precision Instrument 501352
Encompass
The PZMIII offers stereo viewing with ample working distance when used with the long working distance (LWD) option. The long working distance (LWD) option with the 501371 and 501375 on a trinocular microscope shows nearly the same scene on the screen as the viewer sees in the eyepieces.
Catalog No. 50-252-1430