Learn More
Medchemexpress LLC Q-Peptide | 1361235-89-3 | 99.46% | 827.80 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP486610MG
Encompass_Preferred
Q-Peptide is a synthetic peptide derived from angiopoietin-1 (QHREDGS) that has demonstrated the ability to enhance keratinocyte migration. This property suggests its potential role in accelerating wound closure, as observed in a diabetic mouse model. The product is intended for research use only.
  • Increases keratinocyte migration.
  • Improves wound healing in diabetic mouse models.
  • Accelerates wound closure.
  • High purity (99.46%).
  • Available in various quantities for research needs.
Catalog No. 50-003-98421