Learn More
TARGETMOL CHEMICALS INC Q-VD-OPH 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T028210MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Q-VD-OPH (Quinoline-Val-Asp-Difluorophenoxymethylketone) is a pan-caspase inhibitor with potent antiapoptotic properties. purity: 97%
Catalog No. 50-258-1788