Learn More
Selleck Chemical LLC Q-VD-Oph | 99.82% | 1135695-98-5 | 25mg
SDP
Supplier:  Selleck Chemical LLC S731125MG
Encompass_Preferred
Q-VD-Oph (Quinoline-Val-Asp-Difluorophenoxymethylketone) is a potent pan-caspase inhibitor with IC50 ranged from 25 to 400 nM for caspases 1,3,8, and 9. Q-VD-OPh can inhibits HIV infection.
Catalog No. NC2218945