Learn More
Hach Company QAC Reagent 1 Powder Pillows, pk/50
Supplier:  Hach Company 2401066
Encompass
For determination of Quaternary Ammonium Compounds (QAC) by the Direct Binary Complex Method. Hach Method 8337. Range: 0.2 - 5.0 mg/L as CTAB. Pack of 50 Powder Pillows
Catalog No. 50-199-5913