Learn More
Medchemexpress LLC Qingyangshengenin | 84745-94-8 | 500.58 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN13755MG
Encompass_Preferred
Qingyangshengenin is a C21 steroidal aglycone and pregnane glycoside. Isolated from the root of *Cynanchum otophyllum* (Asclepiadaceae), it is also known as NSC 379666. This compound exhibits anti-cancer activity.
  • C21 steroidal aglycone.
  • Pregnane glycoside.
  • Isolated from *Cynanchum otophyllum*.
  • Anti-cancer activity.
Catalog No. 50-004-14707