Learn More
Chemglass Life Sciences Chemglass Quick Connect Set, 3/4 in (M30), 316 Stainless Steel, M30 Male Terminations - For Reactor Setups
SDP
Supplier:  Chemglass Life Sciences CG1969M85
Encompass_Preferred
Quick connect set provides a fast, secure method to connect and disconnect hoses, valves, and accessories on process reactors and manifold systems. Constructed from 316 stainless steel with fluorosilicone seals, the coupling set features shutoffs on both male and female couplers and is rated for -60 to 200° C. Typical ordering is one set (each); two sets are required per reactor setup.
  • 316 stainless steel construction for corrosion resistance.
  • Fluorosilicone seals for chemical and temperature resilience.
  • Shutoff couplers on both ends to prevent fluid loss during disconnection.
  • Wide operating temperature range, -60 to 200° C.
  • Quick, tool-free connection and disconnection for fast system reconfiguration.
  • Designed for use with reactor and manifold systems; two sets commonly required per reactor setup.
Catalog No. NC2895393