Learn More
Selleck Chemical LLC QNZ (EVP4593) 5mg 545380-34-5
SDP
Supplier:  Selleck Chemical LLC S49025MG
Encompass_Preferred
QNZ (EVP4593) shows potent inhibitory activity toward both NF-?B activation and TNF-? production with IC50 of 11 nM and 7 nM in Jurkat T cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4293