Learn More
Cayman Chemical QNZ Inhibitors
SDP
Supplier:  Cayman Chemical 100067345 MG
Encompass_Preferred
An inhibitor of NF-κB activation (IC50 = 11 nM in Jurkat cells); inhibits TNF-α production from LPS-stimulated mouse splenocytes (IC50 = 7 nM)
Catalog No. 50-289-0744