Learn More
Medchemexpress LLC QO-40 | 1259536-70-3 | 98.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130070100MG
Encompass_Preferred
QO-40, a pyrazolo[1,5-a]pyrimidine-7(4H)-one (PPO) derivative, is a KCNQ2/KCNQ3 K+ channels activator with an EC50 of 6.94 μM. It stimulated Ca2+-activated K+ current (IK(Ca)) with an EC50 value of 2.3 μM in pituitary GH3 lactotrophs. It is for research use only and not sold to patients.
  • KCNQ2/KCNQ3 K+ channels activator
  • Stimulates Ca2+-activated K+ current (IK(Ca))
  • Potently augments KCNQ2/3 channels
  • Pyrazolo[1,5-a]pyrimidine-7(4H)-one (PPO) derivative
  • High purity (98.77%)
Catalog No. 50-003-38946