Learn More
Medchemexpress LLC QP5038 | 3031429-88-3 | 99.5% | 361.42 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1610785MG
Encompass_Preferred
QP5038 (compound 28) is an inhibitor of QPCTL with an IC50 value of 3.8 nM. It demonstrates antitumor efficacy.
  • Inhibitor of QPCTL with an IC50 value of 3.8 nM
  • Exhibits antitumor efficacy
Catalog No. 50-003-95199