Learn More
Medchemexpress LLC HY-101092 5mg , QS-21 CAS:141256-04-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010925MG
Encompass_Preferred
HY-101092 5mg QS-21 CAS: 141256-04-4 QS-21, an immunostimulatory saponin, could be used as a potent vaccine adjuvant. QS-21 stimulates Th2 humoral and Th1 cell-mediated immune responses through action on antigen presenting cells (APCs) and T cells. QS-21 can activate the NLRP3 inflammasome with subsequent release of caspase-1 dependent cytokines, IL-1β and IL-18. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1614