Learn More
Sartorius QSol Buffer
Supplier:  Sartorius 91161
Encompass
QSol Buffer Concentrate SolutionUse in Sampling Area Fluid Station for iQue Screener; iQue Screener PLUS; iQue Screener HD; HTFC Use directly in assay media/sample buffer for iQue Screener; iQue Screener HD; HTFC Instrument Compatibility The iQue Qsol Buffer Concentrate Solution is recommended for use with the following flow cytometry equipment: iQue Screener iQue Screener PLUS
Catalog No. 50-289-1715