Learn More
Sentry Air Systems Model 400 Quad Arm Fume Extractor
SDP
Supplier:  Sentry Air Systems SS400FSQ
Encompass
The Quad Arm Fume Extractor supplies powerful fume and particulate control for four operators. This efficient and compact design in ideal for limited space environments that need multiple fume extraction points. The (4) 48 in. (121.92 cm) long flex arms can be easily manipulated and adjusted to source capture chemical fumes and vapors as well as particulate from lab/medical processes. This system is easily portable with a handle and heavy-duty wheels. Schools or training centers can benefit from this economical and efficient multiple operator fume extractor. Multiple filters can be used simultaneously to capture fumes, odors, and particulate. Approximate Airflow: up to 700 CFM (175 CFM per arm)
Catalog No. NC2885522
Add to Cart
Request a Quote