Learn More
Sigma Aldrich Fine Chemicals Biosciences QuadraPureR TU macroporous100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 655422
Encompass
QuadraPure(R) TU is a thiourea-based metal scavenger resin that can be used to prevent metal contamination that occurs during pharmaceutical or fine chemical processing.
Catalog No. 50-290-6686