Learn More
ATCC Quantitative Genomic RNA from Enterovirus D68 (strain US/MO/14-18947)
Supplier: ATCC VR1823DQ
Quantitative Genomic RNA from Enterovirus D68 strain US/MO/14-18947 provides a quantified molecular standard for development, verification, and validation of nucleic-acid-based assays. Use it to generate qPCR standard curves, monitor assay performance, and assess lot-to-lot consistency. For research use only; not for diagnostic or therapeutic applications.
- Product type: molecular standard (genomic RNA).
- Organism/strain: Enterovirus D68 strain US/MO/14-18947.
- Format: frozen; volume 100 μL.
- Concentration: ≥1 x 10^5 copies/μL.
- Storage: -70°C or colder.
- Intended use: research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.