Learn More
ATCC Mycobacterium avium; Strain K-10; genomic DNA
Supplier:  ATCC BAA968D
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR).
Catalog No. NC2196851