Learn More
ATCC Quantitative Genomic DNA from Mycoplasma genitalium strain G37 (ATCC 33530DQ™)
Supplier:  ATCC 33530DQ
Encompass
Quantitative Genomic DNA from Mycoplasmoides genitalium strain G37 (ATCC 33530DQ™) is a molecular standard used for assay development, verification, and validation of molecular-based assays. It supports qPCR calibration for bacterial load determination and is suitable for infectious disease and sexually transmitted infection research. Supplied frozen at ≥1 × 105 copies/μL in 100 μL volume.
Catalog No. 50-238-4260