Learn More
ATCC Quantitative Genomic DNA from; Bacillus atrophaeus; Strain: NRS 1221A
Supplier:  ATCC 9372DQ
Encompass
Quantitative genomic DNA from Bacillus atrophaeus strain NRS 1221A can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3155