Learn More
ATCC Quantitative Genomic DNA from; Morganella morganii; subsp. morganii; Strain: M11
Supplier:  ATCC 25830DQ
Encompass
Quantitative Genomic DNA from Morganella morganii subsp. morganii (ATCC 25830) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-2674