Learn More
ATCC Quantitative Genomic DNA from; Candida tropicalis; Strain: AmMS 227
Supplier:  ATCC 66029DQ
Encompass
Quantitative Genomic DNA from Candida tropicalis can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load.
Catalog No. 50-238-4506
 Restricted Item