Learn More
ATCC Quantitative Genomic DNA from; Gardnerella vaginalis
Supplier:  ATCC 14019DQ
Encompass
Quantitative genomic DNA from Gardnerella vaginalis strain 317 is a whole-genome sequenced product that can be used for assay development, verification, validation, and monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4080