Learn More
ATCC Quantitative Genomic DNA from; Fusobacterium nucleatum; Strain: VPI 4355 [1612A]
Supplier:  ATCC 25586DQ
Encompass
Quantitative Genomic DNA from Fusobacterium nucleatum subsp. nucleatum strain VPI 4355 [1612A] (ATCC 25586) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4691