Learn More
ATCC Quantitative Genomic DNA from; Moraxella catarrhalis; Strain: Ne 11
Supplier:  ATCC 25238DQ
Encompass
Quantitative Genomic DNA from Moraxella catarrhalis strain Ne 11 (ATCC 25238) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4179