Learn More
ATCC Quantitative Genomic DNA from Lactobacillus acidophilus strain Scav (ATCC 4356DQ)
Supplier:  ATCC 4356DQ
Encompass
Quantitative genomic DNA prepared from Lactobacillus acidophilus strain Scav for use as a reference material in molecular assay development, verification, validation, and monitoring. Supplied frozen and intended for laboratory molecular biology applications requiring characterized genomic DNA.
  • Intended for assay development and validation.
  • Provides quantitative genomic DNA from L. acidophilus strain Scav.
  • Stored and shipped frozen at -20°C.
  • Suitable as a reference material or control for molecular methods.
Catalog No. NC3729679