Learn More
ATCC Quantitative Genomic RNA from Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-95DQ™)
Supplier:  ATCC VR95DQ
Encompass
Quantitative Genomic RNA from Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-95DQ™) is a molecular standard used for qPCR assay development, validation, and viral load quantification. It is suitable for respiratory disease and infectious disease research.
Catalog No. 50-238-2128