Learn More
ATCC Quantitative Genomic RNA from; Rubella virus; Strain: M33
Supplier:  ATCC VR315DQ
Encompass
Quantitative Genomic RNA from Rubella virus strain M33 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3801