Learn More
ATCC Quantitative Genomic; RNA from Human; rhinovirus 17; Strain: 33342
Supplier:  ATCC VR1663DQ
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-5072