Learn More
ATCC Quantitative Genomic RNA from Influenza A virus H1N1 strain A/California/07/2009 (H1N1)pdm09 (ATCC VR-1894DQ™)
Supplier:  ATCC VR1894DQ
Encompass
Quantitative Genomic RNA from Influenza A virus H1N1 strain A/California/07/2009 (H1N1)pdm09 (ATCC VR-1894DQ™) is a molecular standard used for qPCR assay development, validation, and viral load quantification. This product is recommended for respiratory disease research and vaccine-related assay workflows.
Catalog No. 50-238-1695