Learn More
ATCC Quantitative Genomic RNA from Human respiratory syncytial virus strain 18537 (ATCC VR-1580DQ™)
Supplier:  ATCC VR1580DQ
Encompass
Quantitative Genomic RNA from Human respiratory syncytial virus strain 18537 (ATCC VR-1580DQ™) is a molecular standard used for qPCR assay development, validation, and respiratory disease research. This product was derived from a virus isolated from a child with acute respiratory illness in Washington, DC.
Catalog No. 50-238-1926