Learn More
QA Bio Inc Ludger BioQuant GPEP A2G2S2 glycopeptide standard
Supplier:  QA Bio Inc BQGPEPA2G2S210U
Encompass
The glycopeptide standard is comprised of an A2G2S2 glycan attached to the asparagine amino acid of a peptide with the sequence Lysine-Valine-Alanine-Asparagine-Lysine-Threonine (KVANKT). •10µg
Catalog No. 50-223-6503