Learn More
ATCC Quantitative Synthetic Human coronavirus HKU1 RNA (ATCC VR-3262SD™)
Supplier:  ATCC VR3262SD
Encompass
Quantitative Synthetic Human coronavirus HKU1 RNA (ATCC VR-3262SD™) includes fragments from the acidic tandem repeat region, growth factor-like protein, NTPase/helicase domain, RNA-dependent RNA polymerase, spike, and nucleocapsid regions. Used for qPCR assay development and respiratory disease research. Manufactured under ISO 13485.
Catalog No. 50-238-2980