Learn More
CTL Scientific Supply Corp QUANTOFIX QUAT
SDP
Supplier:  CTL Scientific Supply Corp 91337
Encompass_Preferred
Test strips for the rapid and reliable determination of quaternary ammonium compounds (QUAT) in solutions. Gradation of scale: 0 - 10 - 25 - 50 - 100 - 250 - 500 - 1000 mg/l 100 strips/box
Catalog No. 50-300-354