Learn More
BANGS LABORATORIES INC Quantum FITC-5 MESF Microsphere Calibration Kit, 2-3 Key Features: Accurate Calibration, 14 mL Total Volume, Functionality: Standardization
Supplier:  BANGS LABORATORIES INC 555PC
Encompass
The Quantum FITC-5 MESF kit is a microsphere calibration standard that enables accurate quantitative fluorescence cytometry. It provides 14 mL of calibrated FITC-labeled microspheres, allowing for enhanced instrument calibration, determination of detection thresholds, and reliable data comparison across instruments over time. This kit is essential for standardizing fluorescence intensity measurements and ensuring precise results in flow cytometry applications.
Catalog No. NC1762433