Learn More
MSC Quantum Storage Floor Pick Rack, Double-Sided Mobile, 2,100 lb Capacity, 30 Yellow Bins
Supplier:  MSC 48521629
Encompass
This double-sided mobile pick rack unit includes shelf bins and is designed for efficient organization and storage in various industrial and laboratory environments. Constructed from steel with a durable powder-coated finish, it offers a robust solution for material handling. The unit is supplied with 30 yellow bins, manufactured from a blend of polyethylene and polypropylene, ensuring a combined load capacity of 2,100 lb.
  • Double-sided mobile pick rack design for versatile use
  • Includes 30 bins for organized storage
  • 2,100 lb load capacity to accommodate heavy items
  • Durable steel construction ensures longevity
  • Powder-coated finish provides added protection
  • Bins made from polyethylene and polypropylene for resilience
  • Features 7 shelves for ample storage space
  • Open shelving style allows for easy access
Catalog No. 50-005-27702