Learn More
Anton Paar QUARTZ CONTROL PLATE, +100°Z/+34°OR for MCP 150
Supplier:  Anton Paar 104463
Encompass_Preferred
Quartz control plate with +100°Z/+34°Optical Rotation used for checking and adjusting the polarimeter scale on the MCP 150.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945
Catalog No. NC1818321