Learn More
CEM Corporation QUARTZ CRUCIBLE 20ML 100/CS
Supplier: CEM Corporation 303040
EM's unique, patented quartz fiber crucibles reduce ashing times by as much as 97% and cool in seconds. The quartz fiber material allows oxygen to circulate around the sample speeding combustion even with our unique crucible caps in place. These unbreakable crucibles withstand temperatures up to 1,000 deg.C and do not need desiccation. Quartz fiber crucible caps can be used with our 50- or 100-ml crucibles. Try them out next time you have to ash a sample that needs to be contained. Ideal for low ash samples and samples with high organic content, they reduce the chance for sample contamination and improve accuracy on ROI and LOI determinations.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.