Learn More
Perkin Elmer US LLC Quartz Hydride C2 Dual Cyclonic Spray Chamber for Optima 2x00/4x00/5x00/7x00 DV/8x00
Supplier:  Perkin Elmer US LLC N0782014
Encompass_Preferred
The dual baffled cyclonic spray chamber provides improved precision and low blanks. Hydride gas port for use with hydrideFAST or with any additional gas.
Catalog No. 50-210-4129