Learn More
Glass Expansion Quartz Torch with 1.5mm Injector for X Series
SDP
Supplier:  Glass Expansion 308081279
Encompass_Preferred
Quartz Torch, single-piece, 1.5 mm id injector, for aqueous and less-volatile organic solvent analysis with Thermo X-Series ICP-MS (3601145 and 3600969). Suitable for routine aqueous and less-volatile organic solvent applications but not inert to samples containing hydrofluoric acid (HF).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7018