Learn More
Perkin Elmer US LLC Quartz Tube Insert
Supplier:  Perkin Elmer US LLC N2411401
Encompass_Preferred
This tube insert is made of pure quartz and meant to be used with the optional oxygen analysis kit for the 2400 Series II CHNS/O Elemental Analyzer.
Catalog No. 50-904-4048