Learn More
Medchemexpress LLC Quemliclustat (AB-680) | 2105904-82-1 | ≥95.0% | 580.8 g/mol | C20H24ClFN4O9P2 | 50MG

Supplier: Medchemexpress LLC HY12528650MG
AB-680 (quemliclustat) is a potent, selective, reversible small-molecule inhibitor of CD73 used in research to block adenosine production and study tumor immunology. It exhibits picomolar potency against human CD73 and is supplied as a high-purity solid for in vitro and preclinical studies.
- Potent, reversible inhibitor of CD73 with Ki ≈ 4.9 pM
- High purity ≥95.0%
- Molecular weight 580.8 g/mol
- Molecular formula C20H24ClFN4O9P2
- Intended for research use only; not for human therapeutic use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.