Learn More
Medchemexpress LLC Quercetin | 117-39-5 | 99.8% | 302.24 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1808510MM/1ML
Encompass_Preferred
Quercetin is a natural flavonoid that acts as a stimulator of recombinant SIRT1 and an inhibitor of PI3K, with IC50 values of 2.4 μM for PI3K γ, 3.0 μM for PI3K δ, and 5.4 μM for PI3K β. This product is intended for research use only.
  • Natural flavonoid
  • Stimulator of recombinant SIRT1
  • PI3K inhibitor with IC50 values for PI3K γ, PI3K δ, and PI3K β
  • Appears in 194 publication(s) in Google Scholar
  • Available in solid and solution forms
Catalog No. 50-003-86874